MilliporeSigma
  • Constituents of Asarum sieboldii with inhibitory activity on lipopolysaccharide (LPS)-induced NO production in BV-2 microglial cells.
Constituents of Asarum sieboldii with inhibitory activity on lipopolysaccharide (LPS)-induced NO production in BV-2 microglial cells.
Chemistry & biodiversity (2008-02-23)
Ah-Reum Han, Hye Jeoung Kim, Minkyu Shin, Moochang Hong, Yang Seok Kim, Hyunsu Bae
ABSTRACT
Bioassay-guided fractionation of the root extract of Asarum sieboldii led to the isolation of the four active compounds (-)-sesamin (1), (2E,4E,8Z,10E)-N-(2-methylpropyl)dodeca-2,4,8,10-tetraenamide (2), kakuol (3), and '3,4,5-trimethoxytoluene' (=1,2,3-trimethoxy-5-methylbenzene; 4), in terms of inhibition of lipopolysaccharide (LPS)-induced nitric oxide (NO) production. Compounds 1-4 showed potent inhibition of NO production, with IC(50) values in the low nanomolar-to-micromolar range. Also isolated were the known compounds methylkakuol (5), '3,5-dimethoxytoluene', safrole, asaricin, methyleugenol, and (-)-asarinin, which were found to be inactive in the above assay. Among the ten known isolates, compounds 1, 2, and 5 were found for the first time in this plant.
Sigma-Aldrich
3,4,5-Trimethoxytoluene, 97%