MilliporeSigma
  • Evaluation of analytical markers characterising different drying methods of parsley leaves (Petroselinum crispum L.).
Evaluation of analytical markers characterising different drying methods of parsley leaves (Petroselinum crispum L.).
Die Pharmazie (2008-01-25)
M Lechtenberg, S Zumdick, C Gerhards, T J Schmidt, A Hensel
ABSTRACT
Drying process of parsley leaves from Petroselinum crispum L. can influence the sensory qualities and aromatic taste of this herbal product. Beside oven-dried material, freeze-dried parsley is getting increasingly into the market. In the course of a search for analytical tools to differentiate oven-dried and lyophilised parsley, a HPLC determination of the 6"-O-malonylapiin to apiin ratio was shown to be a suitable marker system. While the ratio is high for fresh and lyophilised leave material, oven-drying leads to demalonylation and, subsequently, to a low malonylapiin--apiin ratio. Additionally, L*a*b colour measurement can be used for quality control to differentiate between different dried parsley raw materials.