MilliporeSigma
  • Cardiac repolarization during hypoglycaemia and hypoxaemia in healthy males: impact of renin-angiotensin system activity.
Cardiac repolarization during hypoglycaemia and hypoxaemia in healthy males: impact of renin-angiotensin system activity.
Europace : European pacing, arrhythmias, and cardiac electrophysiology : journal of the working groups on cardiac pacing, arrhythmias, and cardiac cellular electrophysiology of the European Society of Cardiology (2008-01-22)
Rikke Due-Andersen, Thomas Høi-Hansen, Niels Vidiendal Olsen, Charlotte Ellen Larroude, Jørgen Kim Kanters, Frans Boomsma, Ulrik Pedersen-Bjergaard, Birger Thorsteinsson
ABSTRACT
Activity in the renin-angiotensin system (RAS) may influence the susceptibility to cardiac arrhythmia. To study the effect of basal RAS activity on cardiac repolarization during myocardial stress induced by hypoglycaemia or hypoxaemia in healthy humans. Ten subjects with high RAS activity and 10 subjects with low RAS activity were studied on three different occasions: (i) hypoglycaemia (nadir P-glucose 2.7 +/- 0.5 mmol/L), (ii) hypoxaemia (nadir pO(2) 5.8 +/- 0.5 kPa), and (iii) normoglycaemic normoxia (control day). QT parameters were registered by Holter monitoring. Hypoglycaemia and hypoxaemia induced QTc prolongation (P < 0.001, both stimuli). The QT/RR slope and the VR increased as a function of hypoglycaemia, but were unaffected by hypoxaemia. Low RAS activity was associated with a steeper QT/RR slope in the recovery phase after both stimuli: hypoglycaemia: P = 0.04; hypoxia: P = 0.03. RAS activity had no impact on QTc [P = 0.48 (hypoglycaemia) and P = 0.40 (hypoxaemia)] or any of the other outcome variables. Basal RAS activity has significant impact on QT dynamics, but not the corrected QT interval, during recovery from hypoglycaemia and hypoxaemia. The impact, however, is modest and more subtle than initially expected. The clinical relevance is unclear.
Sigma-Aldrich
Insulin from porcine pancreas, powder, ≥27 USP units/mg
Insulin (porcine), European Pharmacopoeia (EP) Reference Standard