MilliporeSigma
  • Neuropsychotoxicity of abused drugs: molecular and neural mechanisms of neuropsychotoxicity induced by methamphetamine, 3,4-methylenedioxymethamphetamine (ecstasy), and 5-methoxy-N,N-diisopropyltryptamine (foxy).
Neuropsychotoxicity of abused drugs: molecular and neural mechanisms of neuropsychotoxicity induced by methamphetamine, 3,4-methylenedioxymethamphetamine (ecstasy), and 5-methoxy-N,N-diisopropyltryptamine (foxy).
Journal of pharmacological sciences (2008-01-17)
Takayuki Nakagawa, Shuji Kaneko
ABSTRACT
Psychostimulants including amphetamines and cocaine, opioids including morphine, and some recreational drugs share the ability to cause drug dependence and addiction. Although these drugs of abuse primarily act on distinct molecular targets, such as monoamine transporters or receptors, they finally converge to common neural pathways. Several lines of evidence suggest that their chronic treatment leads to the enhancement of the mesocorticolimbic dopaminergic neurons from the ventral tegmental area (VTA) to the nucleus accumbens (NAc) and the medial prefrontal cortex (mPFC) and leads to abnormal glutamatergic function from the mPFC to the NAc and VTA. The neural adaptation of dopaminergic-glutamatergic system is considered to be critically implicated in neuropsychotoxic effects of these drugs of abuse. In addition, recent studies suggest that the serotonergic neurons from the raphe nuclei to limbic areas modulate the mesocorticolimbic dopaminergic-glutamatergic system and participate in the neuropsychotoxicity. In this review, our recent in vitro studies on the molecular targets and neural adaptation of methamphetamine, 3,4-methylenedioxymethamphetamine (MDMA, "ecstasy"), and 5-methoxy-N,N-diisopropyltryptamine (5-MeO-DiPT, "foxy") using Xenopus oocytes, organotypic slice cultures of the mesocorticolimbic dopaminergic-glutamatergic system, and the raphe serotonergic system are introduced.
Sigma-Aldrich
5-Methoxytryptamine, 97%