MilliporeSigma
  • Ca2+ transport mediated by a synthetic neutral Ca2+ -ionophore in biological membranes.
Ca2+ transport mediated by a synthetic neutral Ca2+ -ionophore in biological membranes.
Biochimica et biophysica acta (1977-11-01)
P Caroni, P Gazzotti, P Vuilleumier, W Simon, E Carafoli
PMID336090
ABSTRACT
The effect of a synthetic neutral ligand on the Ca2+ permeability of several biological membranes has been investigated. The ligand had been previously shown to possess Ca2+ -ionophoric activities in artificial phospholipid membranes. The neutral ionophore is able to transport Ca2+ across the membranes of erythrocytes and sarcoplasmic reticulum, when lipophilic anions such as tetraphenylborate and carbonylcyanide p-trifluoromethoxyphenylhydrazone (FCCP) are present, presumably to facilitate the diffusion of the charged Ca2+ -ionophore complex across the hydrophobic core of the membrane. In mitochondria, the neutral ionophore promotes the active transport of Ca2+ in response to the negative membrane potential generated by respiration, in the presence of the specific inhibitor of the natural carrier ruthenium red.
Product Description
Supelco
Calcium ionophore I, Selectophore, function tested, ≥99.0%