MilliporeSigma
  • Reactions of glyceraldehyde 3-phosphate dehydrogenase sulfhydryl groups with bis-electrophiles produce DNA-protein cross-links but not mutations.
Reactions of glyceraldehyde 3-phosphate dehydrogenase sulfhydryl groups with bis-electrophiles produce DNA-protein cross-links but not mutations.
Chemical research in toxicology (2008-01-01)
Elisabeth M Loecken, F Peter Guengerich
ABSTRACT
The environmental contaminant 1,2-dibromoethane and diepoxybutane, an oxidation product of the important industrial chemical butadiene, are bis-functional electrophiles and are known to be mutagenic and carcinogenic. One mechanism by which bis-electrophiles can exert their toxic effects is through the induction of genotoxic and mutagenic DNA-peptide cross-links. This mechanism has been shown in systems overexpressing the DNA repair protein O6 -alkylguanine DNA-alkyltransferase (AGT) or glutathione S-transferase and involves reactions with nucleophilic cysteine residues. The hypothesis that DNA-protein cross-link formation is a more general mechanism for genotoxicity by bis-electrophiles was investigated by screening nuclear proteins for reactivity with model monofunctional electrophiles. Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) was identified as a candidate because of the nucleophilicity of two cysteine residues (Cys152 and Cys246) in reaction screens with model electrophiles (Dennehy, M. K. et al. (2006) Chem. Res. Toxicol. 19, 20-29). Incubation of GAPDH with bis-electrophiles resulted in inhibition of its catalytic activity, but only at high concentrations of diepoxybutane. In vitro assays indicated DNA-GAPDH cross-link formation in the presence of diepoxybutane, and bis-electrophile reactivity at Cys246 was confirmed using mass spectral analysis. In contrast to AGT, overexpression of human GAPDH in Escherichia coli did not enhance mutagenesis by diepoxybutane. We propose that the lack of mutational enhancement is in part due to the inherently lower reactivity of GAPDH toward bis-electrophiles as well as the reduced DNA binding ability relative to AGT, preventing the in vivo formation of DNA-protein cross-links and enhanced mutagenesis.
Product Description
Supelco
1,2-Dibromoethane solution, certified reference material, 200 μg/mL in methanol
Sigma-Aldrich
1,2-Dibromoethane, 98%
Sigma-Aldrich
1,2-Dibromoethane, purum, ≥98.0% (GC)
Sigma-Aldrich
1,2-Dibromoethane, ≥99%