MilliporeSigma
  • Development of a method for the determination of UV filters in water samples using stir bar sorptive extraction and thermal desorption-gas chromatography-mass spectrometry.
Development of a method for the determination of UV filters in water samples using stir bar sorptive extraction and thermal desorption-gas chromatography-mass spectrometry.
Journal of chromatography. A (2007-12-22)
Rosario Rodil, Monika Moeder
ABSTRACT
Stir bar sorptive extraction (SBSE) in combination with thermal desorption-gas chromatography-mass spectrometry (TD-GC-MS) was applied for the determination of 9 UV filters in water samples. The stir bar coated with polydimethylsiloxane (PDMS) was added to 20 mL of water sample at pH 2 (10% MeOH) and stirred at 1000 rpm for 180 min. Then, the stir bar was subjected to TD-GC-MS. The desorption conditions (desorption temperature and desorption time) and SBSE parameters (ionic strength, pH, presence of organic solvent and time) were optimised using a full factorial design and a Box-Behnken design, respectively. The method shows good linearity (correlation coefficients >0.994) and reproducibility (RSD<16%). The extraction efficiencies were above 63% for all the compounds. Detection limits were between 0.2 and 63 ng/L. The developed method offers the ability to detect several UV filters at ultra-low concentration levels with only 20 mL of sample volume. The effectiveness of the method was tested by analysing real samples such as lake water, river water and treated wastewater. The application of the method allowed reporting the levels of UV filters in environmental water samples.
Sigma-Aldrich
2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 97%
Supelco
Octocrylene, analytical standard
Supelco
Octocrylene, Pharmaceutical Secondary Standard; Certified Reference Material