MilliporeSigma
  • On the mechanism and kinetics of radical reactions of epoxyketones and epoxynitriles induced by titanocene chloride.
On the mechanism and kinetics of radical reactions of epoxyketones and epoxynitriles induced by titanocene chloride.
The Journal of organic chemistry (2007-11-30)
A Fernandez-Mateos, P Herrero Teijón, L Mateos Burón, R Rabanedo Clemente, R Rubio Gonzalez
ABSTRACT
The reactions of a series of epoxynitriles and epoxyketones induced by titanocene chloride have been studied. The kinetics of the decyanogenation of beta,gamma-epoxynitriles with Ti(III) corresponds to a radical reaction (k25 approximately 106 s-1), as demonstrated by competition experiments with H-transfer from 1,4-cyclohexadiene (1,4-CHD) or PhSH or conjugate addition to acrylonitrile. The 5-exo cyclization onto nitrile induced by Ti(III) is a radical reaction (k25 approximately 107 s-1) as seen in competition experiments with H-transfer from PhSH or the titanocene-water complex. The iminyl or alkoxyl radicals generated by 5-exo cyclization onto nitriles or ketones only undergo a reduction with Ti(III). This reaction overwhelms any alternative process, such as tandem cyclization onto alkenes or beta-scission. Iminyl radicals generated by 4-exo cyclizations onto nitriles undergo reduction with Ti(III) and beta-scission reaction in a ratio of 96:4 when the alpha-substituent is CN. Alkoxyl radicals from 4-exo cyclizations onto ketone carbonyls undergo reduction with Ti(III) and beta-scission in a ratio of 60:40 when the alpha-substituent is COOR. In nearly all the reactions studied, the role of Ti(III) is triple: a radical initiator (homolytic cleavage of oxirane), a Lewis acid (coordination to CN or C=O), and a terminator (reduction of iminyl or alkoxyl radicals).
Sigma-Aldrich
Bis(cyclopentadienyl)titanium(IV) dichloride, 97%