MilliporeSigma
Fragrance material review on cinnamyl butyrate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-27)
S P Bhatia, G A Wellington, J Cocchiara, J Lalko, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of cinnamyl butyrate when used as a fragrance ingredient is presented.