MilliporeSigma
  • Potent and selective small molecule NS3 serine protease inhibitors of Hepatitis C virus with dichlorocyclopropylproline as P2 residue.
Potent and selective small molecule NS3 serine protease inhibitors of Hepatitis C virus with dichlorocyclopropylproline as P2 residue.
Bioorganic & medicinal chemistry (2007-11-23)
Kevin X Chen, Bancha Vibulbhan, Weiying Yang, Kuo-Chi Cheng, Rong Liu, John Pichardo, Nancy Butkiewicz, F George Njoroge
ABSTRACT
Starting from a pentapeptide Hepatitis C virus NS3 protease inhibitor, a number of alpha-ketoamide inhibitors based on novel dichlorocyclopropylproline P2 core were synthesized and investigated for their HCV NS3 serine protease activity. The key intermediate 3,4-dichlorocyclopropylproline was obtained through a dichloro carbene insertion to 3,4-dehydroproline. The size of the molecules was reduced significantly through a series of truncations of the initial pentapeptide. By varying P1 side chain in length and size, potency and selectivity were improved. A variety of aliphatic carbamate and urea capping groups were examined. In general, compounds with urea cappings were more potent and selective than their carbamate counterparts. The most potent compound was a tert-butyl urea analog. Variations at P3 position were also investigated. Among the three residues incorporated, tert-leucine was clearly superior, leading to compounds that had excellent enzyme potency and selectivity. The most potent compound achieved cell-based replicon assay EC50 of 40 nM. The most promising compound of all had excellent potency in both enzyme (Ki* = 9 nM) and replicon assays (EC50 = 100 nM). Its bioavailabilities were above 10% in all three animal species (rats, monkeys, and dogs). It has provided a lead for future investigations.
Sigma-Aldrich
N-Butylurea, 99%