MilliporeSigma
  • Functionalization of 6-nitrobenzo[1,3]dioxole with carbonyl compounds via TDAE methodology.
Functionalization of 6-nitrobenzo[1,3]dioxole with carbonyl compounds via TDAE methodology.
Molecules (Basel, Switzerland) (2007-11-17)
Ouassila Amiri-Attou, Thierry Terme, Patrice Vanelle
ABSTRACT
We report herein the synthesis of substituted 2-(6-nitrobenzo[1,3]dioxol-5-yl)-1- aryl ethanols and 2-(6-nitrobenzo[1,3]dioxol-5-yl)-propionic acid ethyl esters from the reaction of 5-chloromethyl-6-nitrobenzo[1,3]dioxole with various aromatic carbonyl and alpha- carbonyl ester derivatives using the tetrakis(dimethylamino)ethylene (TDAE) methodology.
Sigma-Aldrich
2-Dimethylaminoethanol, purified by redistillation, ≥99.5%
Sigma-Aldrich
2-Dimethylaminoethanol, ≥99.5%
Sigma-Aldrich
2-Dimethylaminoethanol, purum, ≥98.0% (GC)