MilliporeSigma
  • Correlation of intracellular diadenosine triphosphate (Ap3A) with apoptosis in Fhit-positive HEK293 cells.
Correlation of intracellular diadenosine triphosphate (Ap3A) with apoptosis in Fhit-positive HEK293 cells.
Cancer letters (2007-11-17)
David I Fisher, Alexander G McLennan
ABSTRACT
The pro-apoptotic Fhit tumor suppressor protein binds and hydrolyses diadenosine triphosphate (Ap3A) and diadenosine tetraphosphate (Ap4A) in vitro. We have measured the level of both these nucleotides in Fhit-positive HEK293 cells exposed to various apoptosis inducers. Cold shock, anti-Fas, cadmium ions and etoposide all increased the basal level of Ap4A of 0.500pmol/10(6)cells by about 50%. However, the corresponding increases in Ap3A from a basal 0.079pmol/10(6)cells correlated closely with the degree of apoptosis produced, up to a maximum of 0.510pmol/10(6)cells with etoposide. These results support the view that Ap3A is the in vivo Fhit ligand and that an inhibition of Fhit activity is a key element in Fhit-mediated apoptosis.
Sigma-Aldrich
Cadmium(II) acetate, anhydrous, 99.995%
Cadmium(II) acetate, SAFC Hitech®, anhydrous, 99.995%
Sigma-Aldrich
Cadmium acetate dihydrate, reagent grade, 98%
Sigma-Aldrich
Cadmium acetate dihydrate, purum p.a., ≥98.0% (KT)