MilliporeSigma
  • Structural studies of two Tinuvin P analogs: 2-(2,4-dimethylphenyl)-2H-benzotriazole and 2-phenyl-2H-benzotriazole.
Structural studies of two Tinuvin P analogs: 2-(2,4-dimethylphenyl)-2H-benzotriazole and 2-phenyl-2H-benzotriazole.
Molecules (Basel, Switzerland) (2007-10-27)
Rosa María Claramunt, Dolores Santa María, Elena Pinilla, M Rosario Torres, José Elguero
ABSTRACT
2-(2,4-Dimethylphenyl)-2H-benzotriazole (1) has been synthesized in a three step procedure starting from 2,4-dimethyl-N-(2-nitrophenyl)benzamide via a 5-(2,4- dimethylphenyl)-1-(2-nitrophenyl)-1H-tetrazole intermediate. Its structure and those of Tinuvin P and 2-phenyl-2H-benzotriazole (5) have been studied by multinuclear NMR (1H-, 13C- and 15N-) in solution and in the solid state. X-ray diffraction analysis of 1 and 5 allowed to us establish the molecular conformation around the single bond connecting the two aromatic systems, in agreement with the conclusions drawn from the NMR study. In the case of 1 ab initio geometry optimization was achieved at the Hartree-Fock HF/6- 31G** and DFT B3LYP/6-31G** levels.
Sigma-Aldrich
2-(2-Hydroxy-5-methylphenyl)benzotriazole, 97%