MilliporeSigma
  • Case report: mixed cholestatic/hepatocellular liver injury induced by the herbicide quizalofop-p-ethyl.
Case report: mixed cholestatic/hepatocellular liver injury induced by the herbicide quizalofop-p-ethyl.
Environmental health perspectives (2007-10-17)
Ioannis S Elefsiniotis, George D Liatsos, Dimitris Stamelakis, Antonios Moulakakis
ABSTRACT
Quizalofop-p-ethyl is an often applied, slightly toxic herbicide for which no severe toxicity has been reported in humans. We present the case of a farmer exposed to quizalofop-p-ethyl who presented with obstructive cholestasis. A complete workup disclosed no other cause of liver pathology, but liver biopsy established drug-induced hepatotoxicity. The patient was treated with ursodeoxycholic acid and prednisolone, and was recovered fully 70 days after his exposure to the herbicide. The patient was followed for the next 9 months. Quizalofop-p-ethyl can induce a mixed cholestatic/hepatocellular liver injury. We discuss possible mechanisms implicated in liver injury after exposure to quizalofop-p-ethyl. In patients presenting with mixed cholestatic/ hepatocellular liver injury, occupational exposure to quizalofop-p-ethyl in the course of agricultural use should be investigated.
Product Description
Supelco
Quizalofop-P-ethyl, PESTANAL®, analytical standard
Supelco
Quizalofop-ethyl, PESTANAL®, analytical standard