MilliporeSigma
  • Traceless liquid-phase synthesis of biphenyls and terphenyls using pentaerythritol as a tetrapodal soluble support.
Traceless liquid-phase synthesis of biphenyls and terphenyls using pentaerythritol as a tetrapodal soluble support.
Journal of combinatorial chemistry (2007-09-29)
Chul-Bae Kim, Chul-Hee Cho, Chang Keun Kim, Kwangyong Park
ABSTRACT
Application of a novel sulfonate-based traceless multifunctional linker system using pentaerythritol as a tetrapodal soluble support was demonstrated using liquid-phase parallel and combinatorial preparation of biphenyl and terphenyl compounds. Nickel-catalyzed reactions of pentaerythritol tetrakis(arenesulfonate)s with arylmagnesium bromides generated the desired products in sufficient yields through reductive cleavage/cross-coupling of the C-S bond. Homogeneous pentaerythritol-supported reactions could be accomplished using less nucleophile with shorter reaction periods than could the corresponding heterogeneous polymer-supported reactions. This liquid-phase approach using a small polyfunctionalized support combines advantages of solution-phase and solid-phase syntheses by allowing high reactivity, high atom economy, simple isolation, and real-time monitoring of the reaction progress.
Sigma-Aldrich
Pentaerythritol, 98%
Sigma-Aldrich
Pentaerythritol, 99%