MilliporeSigma
  • Regioselective formation of 6-O-acylsucroses and 6,3'-di-O-acylsucroses via the stannylene acetal method.
Regioselective formation of 6-O-acylsucroses and 6,3'-di-O-acylsucroses via the stannylene acetal method.
Carbohydrate research (2007-09-26)
Qinghui Wang, Shufen Zhang, Jinzong Yang
ABSTRACT
Regioselective formation of 6-O-acylsucroses and 6,3'-di-O-acylsucroses in one pot with good yields was achieved for the first time by a typical acylation method of sucrose via its dibutylstannylene acetal. Pure monoesters at OH-6 and diesters at OH-6,3' obtained by these procedures were readily isolated by simple column chromatography, thus overcoming the main difficulties associated with regioselectivity, efficiency, and isolation techniques for the practical preparation. Explanations for the regioselectivities observed during this stannylene acetal-mediated reaction were also proposed based on the structures of the stannylene acetal in solution and the intramolecular migration of stannylenes.
Sigma-Aldrich
Dibutyltin(IV) oxide, 98%