MilliporeSigma
  • Formation of an exciplex with mixed ligands in the mercury-photosensitized luminescence of alcohol-amine mixtures in the liquid phase.
Formation of an exciplex with mixed ligands in the mercury-photosensitized luminescence of alcohol-amine mixtures in the liquid phase.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-09-07)
Shunzo Yamamoto, Michiyo Uno, Yoshimi Sueishi
ABSTRACT
The liquid-phase mercury-photosensitized luminescence of tert-butyl alcohol (TL)-tert-butylamine (TM) mixtures has been investigated by a steady-state illumination method over a wide range of substrate concentrations. The emission bands from exciplexes (HgTL* and HgTM*) between an excited mercury atom and an alcohol or an amine molecule were observed at about 330 nm and 370 nm, respectively, in TL and TM solutions in cyclohexane. Two other bands appeared at 405 nm and 455 nm for TM at high concentrations. These bands were previously assigned to two types of 1:2 exciplexes (HgTM(2)* and HgTM(2)**). In TL-TM mixed solutions, a new band appeared at about 400 nm. The intensity of this band increased with increasing concentrations of TL and TM. This band was attributed to an exciplex with mixed ligands (HgTLTM*). This band was observed for the first time in this study. The energized intermediate, (HgTLTM*)(not equal), formed between HgAL* and AM can be effectively stabilized by collisions with solvent molecules in solution, while it decomposes to HgAM* and AL in the gas phase. The results for TL-TM mixtures can be explained by the proposed reaction mechanism.
Sigma-Aldrich
tert-Butylamine, 98%
Sigma-Aldrich
tert-Butylamine, ≥99.5%
Sigma-Aldrich
tert-Butylamine hydrochloride, ≥98.0% (AT)