MilliporeSigma
  • Protein engineering of nitrilase for chemoenzymatic production of glycolic acid.
Protein engineering of nitrilase for chemoenzymatic production of glycolic acid.
Biotechnology and bioengineering (2007-09-06)
Shijun Wu, Arthur J Fogiel, Kelly L Petrillo, Raymond E Jackson, Kimberley N Parker, Robert Dicosimo, Arie Ben-Bassat, Daniel P O'Keefe, Mark S Payne
ABSTRACT
A key step in a chemoenzymatic process for the production of high-purity glycolic acid (GLA) is the enzymatic conversion of glycolonitrile (GLN) to ammonium glycolate using a nitrilase derived from Acidovorax facilis 72W. Protein engineering and over-expression of this nitrilase, combined with optimized fermentation of an E. coli transformant were used to increase the enzyme-specific activity up to 15-fold and the biocatalyst-specific activity up to 125-fold. These improvements enabled achievement of the desired volumetric productivity and biocatalyst productivity for the conversion of GLN to ammonium glycolate.
Sigma-Aldrich
Glycolic acid nitrile solution, ~70% in H2O