MilliporeSigma
  • Ruthenium(II) complexes of 2-benzoylpyridine-derived thiosemicarbazones with cytotoxic activity against human tumor cell lines.
Ruthenium(II) complexes of 2-benzoylpyridine-derived thiosemicarbazones with cytotoxic activity against human tumor cell lines.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-08-21)
Angelica E Graminha, Cláudia Rodrigues, Alzir A Batista, Letícia R Teixeira, Elaine S Fagundes, Heloisa Beraldo
ABSTRACT
Reaction of [RuCl(3)(dppb)H(2)O] (dppb=1,4 bis(diphenylphospine)butane) with 2-benzoylpyridine thiosemicarbazone (H2Bz4DH) and its N(4)-methyl (H2Bz4M) and N(4)-phehyl (H2Bz4Ph) derivatives gave [RuCl(dppb)(H2Bz4DH)]Cl (1), [RuCl(dppb)(H2Bz4M)]Cl (2) and [RuCl(dppb)(H2Bz4Ph)]Cl (3). The cytotoxic activity of the studied compounds was tested against the MCF-7, TK-10 and UACC-62 human tumor cell lines. The precursor [RuCl(3)(dppb)H(2)O] exhibits cytocidal activity against the tree cell lines. H2BzDH, H2Bz4M, and [RuCl(dppb)(H2Bz4M)]Cl (2) show a selective cytocidal effect against the UACC-62 cell line which makes them the most promising compounds.
Sigma-Aldrich
2-Benzylpyridine, 98%