MilliporeSigma
  • Simultaneous determination of rivanol and mifepristone in human plasma by a HPLC-UV method with solid-phase extraction.
Simultaneous determination of rivanol and mifepristone in human plasma by a HPLC-UV method with solid-phase extraction.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2007-08-11)
Zhiyong Guo, Danyi Wei, Gengxin Yin, Sui Wang, Shasha Zhao, Yun Chu, Jinxia Zhai
ABSTRACT
A HPLC method with UV detection was developed and validated for the simultaneous determination of rivanol and mifepristone in human plasma. Norethisterone was used as the internal standard. Separation was performed by a C18 reversed-phase column maintained at 20 degrees C. The mobile phase was a mixture of methanol-acetonitrile-0.05% sodium dodecylsulfonate in a 0.05 M phosphate buffer with the pH adjusted to 3.0 (30:30:40, v/v/v) at a flow rate of 0.8 ml/min. Dual wavelength mode was used, with mifepristone monitored at UV 302 nm, while rivanol and norethisterone at 272 nm. A reliable biological sample pre-treatment procedure by means of solid-phase extraction was used, which allowed to obtain good extraction efficiency (>93%) for both of the analytes and the internal standard. The calibration curves were both linear with the correlation coefficient r equal to 0.9999. For rivanol, the assay gave CV% values for precision always lower than 7.8% and mean accuracy values higher than 95.3%. As to mifepristone, precision was always lower than 10.1% and mean accuracy values were higher than 93.8%. The limit of detection for the assay of rivanol and mifepristone was 1.1 and 3 ng/ml, respectively. The method is simple, sensitive and accurate, and allow for simultaneous determination of nanogram levels of rivanol and mifepristone in human plasma. It could be applied to assess the plasma level of rivanol and mifepristone in women undergoing polypharmacy with the two drugs.
Sigma-Aldrich
6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate, 99%