MilliporeSigma
  • Poststaining grids for transmission electron microscopy: conventional and alternative protocols.
Poststaining grids for transmission electron microscopy: conventional and alternative protocols.
Methods in molecular biology (Clifton, N.J.) (2007-07-28)
E Ann Ellis
ABSTRACT
Poststaining ultrathin sections on grids is an essential process in preparing specimens for examination using transmission electron microscopy. This process commonly consists of staining in aqueous solutions of uranyl acetate followed by lead citrate and can result in good consistent quality staining if certain precautions are taken. This chapter covers conventional and alternative methods for poststaining ultrathin sections, including microwave-assisted procedures and corrective measures when problems are encountered.
Sigma-Aldrich
Lead(II) carbonate, ACS reagent