MilliporeSigma
Room-temperature copper-catalyzed alpha-arylation of malonates.
Organic letters (2007-07-20)
Sau Fan Yip, Hong Yee Cheung, Zhongyuan Zhou, Fuk Yee Kwong
ABSTRACT
An effective method in targeting alpha-aryl malonates is reported. In the presence of a catalytic amount of 2-picolinic acid and CuI, the coupling of aryl iodides with diethyl malonate proceeds smoothly even at room temperature. The high levels of functional group compatibility and exceptionally mild reaction conditions offer this an attractive protocol in accessing a variety of arylated malonates.
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl malonate, ≥98%, FG