MilliporeSigma
[Purely natural: phototoxic dermatitis].
MMW Fortschritte der Medizin (2007-07-11)
E Knott, H Hofmann
ABSTRACT
The local application of undiluted bergamot oil in association with simultaneous exposure to sunlight brought about a phototoxic reaction of the skin. Despite successful treatment of the vesicles, Hyperpigmentation and stress-related erythema persisted.
Sigma-Aldrich
Bergamot oil