MilliporeSigma
  • Thermodynamic determination of the binding constants of angiotensin-converting enzyme inhibitors by a displacement method.
Thermodynamic determination of the binding constants of angiotensin-converting enzyme inhibitors by a displacement method.
FEBS letters (2007-07-10)
Montserrat Andújar-Sánchez, Vicente Jara-Pérez, Ana Cámara-Artigas
ABSTRACT
Somatic angiotensin I-converting enzyme (s-ACE) plays a central role in blood pressure regulation and has been the target of most antihypertensive drugs. A displacement isothermal titration calorimetry method has been used to accurately determine the binding constant of three strong s-ACE inhibitors. Under the experimental conditions studied in this work, the relative potency of the inhibitors was determined to be enalaprilat>lisinopril>captopril. We analyze the thermodynamic behaviour of the binding process using the new structural information provided by the ACE structures, as well as the conformational changes that occur upon binding.
Sigma-Aldrich
Asp-Phe, 96%