MilliporeSigma
  • Pharmacokinetics of florfenicol and its metabolite, florfenicol amine, in dogs.
Pharmacokinetics of florfenicol and its metabolite, florfenicol amine, in dogs.
Research in veterinary science (2007-06-16)
Byung-Kwon Park, Jong-Hwan Lim, Myoung-Seok Kim, Youn-Hwan Hwang, Hyo-In Yun
ABSTRACT
A study on the bioavailability and pharmacokinetics of florfenicol was conducted in six healthy dogs following a single intravenous (i.v.) or oral (p.o.) dose of 20 mg kg(-1) body weight (b.w.). Florfenicol concentrations in serum were determined by a high-performance liquid chromatography/mass spectrometry. Plasma concentration-time data after p.o. or i.v. administration were analyzed by a non-compartmental analysis. Following i.v. injection, the total body clearance was 1.03 (0.49) L kg(-1)h(-1) and the volume of distribution at steady-state was 1.45 (0.82) L kg(-1). Florfenicol was rapidly distributed and eliminated following i.v. injection with 1.11 (0.94)h of the elimination half-life. After oral administration, the calculated mean C(max) values (6.18 microg ml(-1)) were reached at 0.94 h in dogs. The elimination half-life of florfenicol was 1.24 (0.64) h and the absolute bioavailability (F) was achieved 95.43 (11.60)% after oral administration of florfenicol. Florfenicol amine, the major metabolite of florfenicol, was detected in all dogs after i.v. and p.o. administrations.
Product Description
Supelco
Florfenicol amine, VETRANAL®, analytical standard