MilliporeSigma
  • Temperature and pH effect on the microbial reductive transformation of pentachloronitrobenzene.
Temperature and pH effect on the microbial reductive transformation of pentachloronitrobenzene.
Journal of agricultural and food chemistry (2007-06-08)
Didem Okutman Tas, Spyros G Pavlostathis
ABSTRACT
The effect of pH and temperature on the microbial reductive transformation of pentachloronitrobenzene (PCNB), an organochlorine fungicide, was investigated with a mixed fermentative/methanogenic culture developed from a contaminated estuarine sediment. Culture series were incubated at a temperature range from 4 to 45 degrees C at pH 6.9+/-0.1 and at a pH range from 2.7+/-0.1 to 7.6+/-0.1 at 22 degrees C. Significant differences were observed in terms of biotransformation rate, extent, and products as a function of temperature. Incubation at different pH values resulted in differences in biotransformation rate and extent, but not in terms of products formed. PCNB (3 microM) was transformed to pentachloroaniline (PCA) in all culture series. However, sequential dechlorination of PCA was observed only at a temperature range from 4 to 35 degrees C and at a pH range from 6.2+/-0.1 to 7.6+/-0.1. The highest PCA dechlorination rate was observed at 22 degrees C and at pH 7.6+/-0.1. The effect of temperature on the PCA dechlorination rate was modeled using an Arrhenius relationship, which accounts for both enzyme activation and deactivation. The dechlorination of PCA and chlorinated aniline intermediates was simulated using a branched-chain Michaelis-Menten model, and kinetic constants were determined.
Sigma-Aldrich
Pentachloronitrobenzene, ≥94%
Supelco
Pentachloronitrobenzene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Quintozene, PESTANAL®, analytical standard