MilliporeSigma
  • Treatment of 1,10-phenanthroline laboratory wastewater using the solar photo-Fenton process.
Treatment of 1,10-phenanthroline laboratory wastewater using the solar photo-Fenton process.
Journal of hazardous materials (2007-05-29)
Milady Renata Apolinário da Silva, Alam Gustavo Trovó, Raquel Fernandes Pupo Nogueira
ABSTRACT
The red Fe(2+)-phenanthroline complex is the basis of a classical spectrophotometric method for determination of iron. Due to the toxicity of this complexing agent, direct disposal of the wastewaters generated in analytical laboratories is not environmentally safe. This work evaluates the use of the solar photo-Fenton process for the treatment of laboratory wastewaters containing phenanthroline. Firstly, the degradation of phenanthroline in water was evaluated at two concentration levels (0.1 and 0.01%, w/v) and the efficiencies of degradation using ferrioxalate (FeOx) and ferric nitrate were compared. The 0.01% w/v solution presented much higher mineralization, achieving 82% after 30min of solar irradiation with both iron sources. The solar photo-Fenton treatment of laboratory wastewater containing, in addition to phenanthroline, other organic compounds such as herbicides and 4-chlorophenol, equivalent to 4,500mgL(-1) total organic carbon (TOC) resulted in total degradation of phenanthroline and 25% TOC removal after 150min, in the presence of either FeOx or ferric nitrate. A ratio of 1:10 dilution of the residue increased mineralization in the presence of ferrioxalate, achieving 38% TOC removal after 120min, while use of ferric nitrate resulted in only 6% mineralization over the same period.
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ACS reagent, ≥98%