MilliporeSigma
Electron induced ortho-meta isomerization of single molecules.
Physical review letters (2007-05-16)
V Simic-Milosevic, M Mehlhorn, K-H Rieder, J Meyer, K Morgenstern
ABSTRACT
A scanning tunneling microscope operating at 5 K is used to induce the isomerization of single chloronitrobenzene molecules on Cu(111) and verify the reaction. The threshold voltage of (227+/-7) mV for this reaction is explained based on electron-induced vibrational heating. We propose that the isomerization is initiated by simultaneous excitation of two vibrational molecular modes via inelastically tunneling electrons. This excitation results in a shift of the distribution probability of chlorine and hydrogen positions, which facilitates their mutual exchange.
Sigma-Aldrich
1-Chloro-2-nitrobenzene, 99%