MilliporeSigma
  • Reaction of perfluorocyclopentene with various carbon nucleophiles--heteroaromatic lithium reagents, enolate and phosphonium ylide.
Reaction of perfluorocyclopentene with various carbon nucleophiles--heteroaromatic lithium reagents, enolate and phosphonium ylide.
Organic & biomolecular chemistry (2007-04-28)
Shigeyuki Yamada, Emi Ishii, Tsutomu Konno, Takashi Ishihara
ABSTRACT
The addition of heteroaromatic lithium reagents 2 to a THF solution of perfluorocyclopentene (1) provided preferentially the corresponding monosubstituted products 5, while the addition of 1 to 2 effectively gave the 1,2-disubstituted products 6 in good to excellent yields. The reaction of 1 with sodiomalonate 3 or phosphonium ylide 4 also proceeded smoothly to form the 1,3-disubstituted product 8 or 10 in high yield, respectively.
Sigma-Aldrich
Diethyl malonate, ≥98%, FG
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%