MilliporeSigma
  • The Kulinkovich reaction in the synthesis of constrained n,n-dialkyl neurotransmitter analogues.
The Kulinkovich reaction in the synthesis of constrained n,n-dialkyl neurotransmitter analogues.
Organic letters (2007-04-24)
Catherine A Faler, Madeleine M Joullié
ABSTRACT
An intermolecular Ti(IV)-mediated cyclopropanation reaction has been used to synthesize substituted 2-phenylcyclopropylamines and constrained analogues of the neurotransmitters histamine and tryptamine. Many hydroxy- and methoxy-substituted phenylcyclopropylamines are known to inhibit monoamine oxidase and have been shown to mimic hallucinogens. These compounds were made in 1 to 5 steps from readily available starting materials.
Sigma-Aldrich
Cyclopropylamine, 98%