MilliporeSigma
  • [Astragalus membranaceus promote expression of insulin-like growth factor 1 in rat model of olivo-cerebellar degeneration].
[Astragalus membranaceus promote expression of insulin-like growth factor 1 in rat model of olivo-cerebellar degeneration].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-04-17)
Hui-ling Wu, Hui-jun Zhou
ABSTRACT
To observe the effect of Astragalus membranaceus (AM) on insulin-like growth factor 1 (IGF-1) expression in a rat model of olivo-cerebellar degeneration and assess the neuroprotective actions of AM meanwhile. Rats model of olivo-cerebellar degeneration was established by using 3-acetylpyridine. The effect of AM on the expression of Calbindin D-28K in inferior olive (IO) neurons by immunohistochemistry, the serum IGF-1 level by Elisa, the IGF-1 mRNA level in the cerebellum by RT-PCR were detected respectively. AM effectively improve the serum IGF-1 level, Cerebellar IGF-1 mRNA level and the survival of the 10 neurons in a rat model of olivo-cerebellar degeneration, even at a lower dose (9 g x kg(-1)), and the effect was in a dose-dependent manner. AM could effectively upregulate the IGF-1 expression in the rat model of olivo-cerebellar degeneration, and have neuroprotective effect on IO neurons.
Sigma-Aldrich
3-Acetylpyridine, 98%
Sigma-Aldrich
3-Acetylpyridine, ≥98%, FG