MilliporeSigma
  • Influence of a novel radiopacifier on the properties of an injectable calcium phosphate cement.
Influence of a novel radiopacifier on the properties of an injectable calcium phosphate cement.
Acta biomaterialia (2007-04-07)
Xiupeng Wang, Jiandong Ye, Yingjun Wang
ABSTRACT
An injectable calcium phosphate cement (CPC) with excellent radiopacity was proposed by introducing a novel radiopacifier, strontium carbonate, into the powder phase of CPC. The results showed that the cement showed improved radiopacity even when the content of strontium carbonate was only 8 or 12wt.%. The addition of 8 or 12wt.% strontium carbonate clearly improved the injectability and compressive strength of the cement. Furthermore, the addition of strontium carbonate influenced the pore distribution in the cement. An injectable CPC containing 8 or 12wt.% strontium carbonate has the potential for use in procedures such as vertebroplasty and kyphoplasty.
Sigma-Aldrich
Strontium carbonate, ≥99.9% trace metals basis
Sigma-Aldrich
Strontium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Strontium carbonate, ≥98%