MilliporeSigma
  • Electrokinetic removal of 2,6-dichlorophenol and diuron from kaolinite and humic acid-clay system.
Electrokinetic removal of 2,6-dichlorophenol and diuron from kaolinite and humic acid-clay system.
Journal of hazardous materials (2007-04-07)
Anna Maria Polcaro, Annalisa Vacca, Michele Mascia, Simonetta Palmas
ABSTRACT
This paper presents the results of a study on the electrokinetic treatment of kaolinite and humic acid kaolinite complexes spiked with 2,6-dichlorophenol or 3-(3,4-dichlorophenyl)-1,1-dimethylurea (diuron). In particular, the attention was paid to the interaction between solid surface and dissolved organics: the effects of contaminant sorption as well as the physicochemical reactions on the efficiency of electrokinetic remediation were investigated. Using a 3V/cm voltage gradient, approximately 90% of diuron was removed from kaolinite after one water pore volume was collected in the cathode reservoir, but much lower efficiency was obtained in the electrokinetic removal of this compound from humic acid-coated kaolinite. The results also showed that partial degradation of the contaminant occurred during electrokinetic treatment of kaolinite clay spiked with 2,6-dichlorophenol: the contamination in the clay could be remediated by the combination of electrokinetic extraction and electrochemical reactions.
Sigma-Aldrich
2,6-Dichlorophenol, 99%
Supelco
2,6-Dichlorophenol, PESTANAL®, analytical standard