MilliporeSigma
  • Non-fouling microfluidic chip produced by radio frequency tetraglyme plasma deposition.
Non-fouling microfluidic chip produced by radio frequency tetraglyme plasma deposition.
Lab on a chip (2007-03-29)
Malinda Salim, Gautam Mishra, Gregory J S Fowler, Brian O'sullivan, Phillip C Wright, Sally L McArthur
ABSTRACT
This Technical Note presents the direct surface modification of a glass/PTFE hybrid microfluidic chip, via radio frequency glow discharge plasma polymerisation of tetraethlylene glycol dimethylether (tetraglyme), to produce hydrophilic, non-fouling, PEO-like surfaces. We use several techniques including X-ray photoelectron spectroscopy (XPS), direct enzyme-linked immunosorbent assays (ELISA) and immunofluorescent imaging to investigate the channel coatings. Our results indicate the successful deposition of a PEO-like coating onto microchannel surfaces that has both solution and shelf stability (>3 months) and is capable of preventing fibrinogen adsorption to the microchannel surfaces.
Sigma-Aldrich
Tetraethylene glycol dimethyl ether, ≥99%