MilliporeSigma
  • Ultra-trace determination of beryllium in occupational hygiene samples by ammonium bifluoride extraction and fluorescence detection using hydroxybenzoquinoline sulfonate.
Ultra-trace determination of beryllium in occupational hygiene samples by ammonium bifluoride extraction and fluorescence detection using hydroxybenzoquinoline sulfonate.
Analytica chimica acta (2007-03-28)
Kevin Ashley, Anoop Agrawal, John Cronin, Juan Tonazzi, T Mark McCleskey, Anthony K Burrell, Deborah S Ehler
ABSTRACT
A highly sensitive molecular fluorescence method for measuring ultra-trace levels of beryllium has been previously described. The method entails extraction of beryllium workplace samples by 1% ammonium bifluoride (NH(4)HF(2), aqueous), followed by fluorescence detection using hydroxybenzoquinoline sulfonate (HBQS). In this work, modification of the existing procedure resulted in a significant improvement in detection power, thereby enabling ultra-trace determination of beryllium in air filter and surface wipe samples. Such low detection limits may be necessary in view of expected decreases in applicable occupational exposure limits (OELs) for beryllium. Attributes of the modified NH(4)HF(2) extraction/HBQS fluorescence method include method detection limits (MDLs) of <0.8 ng to approximately 2 ng Be per sample (depending on the fluorometer used), quantitative recoveries from beryllium oxide, a dynamic range of several orders of magnitude, and freedom from interferences. Other key advantages of the technique are field portability, relatively low cost, and high sample throughput. The method performance compares favorably with that of inductively coupled plasma-mass spectrometry (ICP-MS).
Sigma-Aldrich
Beryllium oxide, 99.98% trace metals basis