MilliporeSigma
  • Synthesis, thermal and spectral studies of first-row transition metal complexes with Girard P reagent-based ligand.
Synthesis, thermal and spectral studies of first-row transition metal complexes with Girard P reagent-based ligand.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-02-24)
Usama El-Ayaan, I M Kenawy, Y G Abu El-Reash
ABSTRACT
A new series of first-row transition metal complexes with 1-acetylpyridinium chloride-4-benzoyl thiosemicarbazide (H2GPBzIT) have been prepared and characterized by elemental analysis, spectroscopic and magnetic measurements. The proton-ligand ionization constants were determined potentiometrically using Irving-Rossotti technique. The stability constants of complexes were also calculated and were found in agreement with the sequence of stability constants of Irving and Williams. Thermal stability and degradation kinetics have been measured using thermogravimetric analyzer. Kinetic parameters were obtained for each stage of thermal degradation of complexes using Coats-Redfern method.
Sigma-Aldrich
Girard′s reagent T, 98%