MilliporeSigma
  • l-Methionine sulfoximine, but not phosphinothricin, is a substrate for an acetyltransferase (gene PA4866) from Pseudomonas aeruginosa: structural and functional studies.
l-Methionine sulfoximine, but not phosphinothricin, is a substrate for an acetyltransferase (gene PA4866) from Pseudomonas aeruginosa: structural and functional studies.
Biochemistry (2007-01-27)
Anna M Davies, Renée Tata, Rebecca L Beavil, Brian J Sutton, Paul R Brown
ABSTRACT
The gene PA4866 from Pseudomonas aeruginosa is documented in the Pseudomonas genome database as encoding a 172 amino acid hypothetical acetyltransferase. We and others have described the 3D structure of this protein (termed pita) [Davies et al. (2005) Proteins: Struct., Funct., Bioinf. 61, 677-679; Nocek et al., unpublished results], and structures have also been reported for homologues from Agrobacterium tumefaciens (Rajashankar et al., unpublished results) and Bacillus subtilis [Badger et al. (2005) Proteins: Struct., Funct., Bioinf. 60, 787-796]. Pita homologues are found in a large number of bacterial genomes, and while the majority of these have been assigned putative phosphinothricin acetyltransferase activity, their true function is unknown. In this paper we report that pita has no activity toward phosphinothricin. Instead, we demonstrate that pita acts as an acetyltransferase using the glutamate analogues l-methionine sulfoximine and l-methionine sulfone as substrates, with Km(app) values of 1.3 +/- 0.21 and 1.3 +/- 0.13 mM and kcat(app) values of 505 +/- 43 and 610 +/- 23 s-1 for l-methionine sulfoximine and l-methionine sulfone, respectively. A high-resolution (1.55 A) crystal structure of pita in complex with one of these substrates (l-methionine sulfoximine) has been solved, revealing the mode of its interaction with the enzyme. Comparison with the apoenzyme structure has also revealed how certain active site residues undergo a conformational change upon substrate binding. To investigate the role of pita in P. aeruginosa, a mutant strain, Depp4, in which pita was inactivated through an in-frame deletion, was constructed by allelic exchange. Growth of strain Depp4 in the absence of glutamine was inhibited by l-methionine sulfoximine, suggesting a role for pita in protecting glutamine synthetase from inhibition.
Sigma-Aldrich
L-Methionine sulfone
Sigma-Aldrich
DL-Methionine sulfone, ≥99.0% (NT)