MilliporeSigma
  • Development of three parallel cytochrome P450 enzyme affinity detection systems coupled on-line to gradient high-performance liquid chromatography.
Development of three parallel cytochrome P450 enzyme affinity detection systems coupled on-line to gradient high-performance liquid chromatography.
Drug metabolism and disposition: the biological fate of chemicals (2007-01-26)
Jeroen Kool, Sebastiaan M van Liempd, Huub van Rossum, Danny A van Elswijk, Hubertus Irth, Jan N M Commandeur, Nico P E Vermeulen
ABSTRACT
A high resolution screening (HRS) technology is described, in which gradient high-performance liquid chromatography (HPLC) is connected on-line to three parallel placed bioaffinity detection systems containing mammalian cytochromes P450 (P450s). The three so-called enzyme affinity detection (EAD) systems contained, respectively, liver microsomes from rats induced by beta-naphthoflavone (CYP1A activity), phenobarbital (CYP2B activity), and dexamethasone (CYP3A activity). Each P450-EAD system was optimized for enzyme, substrate, and organic modifier (isopropyl alcohol, methanol, and acetonitrile) in flow injection analysis mode. Characteristic P450 ligands were used to validate the P450-EAD systems. IC(50) values of the ligands were measured and found to be similar to those obtained with conventional microtiter plate reader assays. Detection limits (n = 3; signal-to-noise ratio = 3) of potent inhibitors ranged from 1 to 3 pmol for CYP1A activity, 4 to 17 pmol for CYP2B activity, and 4 to 15 pmol for CYP3A activity. The three optimized P450-EAD systems were subsequently coupled to gradient HPLC and used to screen compound mixtures for individual ligands. Finally, to increase analysis efficiency, a HRS system was constructed in which all three P450-EAD systems were coupled on-line and in parallel to gradient HPLC. The triple parallelized P450-EAD system was shown to enable rapid profiling of individual components in complex mixtures for inhibitory activity to three different P450s.
Sigma-Aldrich
Resorufin pentyl ether