MilliporeSigma
  • [The sources of the innervation of the sphincter of the esophagogastric junction in rats].
[The sources of the innervation of the sphincter of the esophagogastric junction in rats].
Arkhiv anatomii, gistologii i embriologii (1991-05-01)
L L Kolesnikov
ABSTRACT
The investigation has been performed by means of the luminescent microscopical method. The retrograde axonal transport of the fluorescent marker primuline has demonstrated that a definite amount of labelled cells are observed in the celiac plexus, in nodes of the thoracic part of the sympathetic trunk (predominantly in Th6-Th8). Innervation of the EGP sphincter is mainly performed from the sympathetic trunk nodes (Th6-Th8) and from the celiac plexus.
Product Description
Supelco
Malvidin-3-galactoside chloride, analytical standard