MilliporeSigma
  • Determination of UV filters and antimicrobial agents in environmental water samples.
Determination of UV filters and antimicrobial agents in environmental water samples.
Analytical and bioanalytical chemistry (2006-12-01)
Petra Cuderman, Ester Heath
ABSTRACT
Although there is increasing concern about residues from personal care products entering the aquatic environment and their potential to accumulate to levels that pose a health threat to humans and wildlife, we still know little about the extent and magnitude of their presence in the aquatic environment. In this study we describe a procedure for isolation, and subsequent determination, of compounds commonly added to personal care products. The compounds of interest include UV filters with the commercial name Eusolex (homosalate, 4-methylbenzylidenecamphor, benzophenone-3, octocrylene, butylmethoxydibenzoylmethane, ethylhexyl methoxycinnamate) and two common anti-microbial agents, clorophene and triclosan. Water samples were filtered, acidified, and extracted by use of solid-phase extraction. Extracted compounds were then derivatised before analysis by gas chromatography-mass spectroscopy. By use of our method we obtained limits of detection of 13-266 ng L(-1) for UV filters, and 10-186 ng L(-1) for triclosan and clorophene. Recoveries were 82-98% for deionised water and 50-98% for natural water (seawater, pool water, lake water, and river water). Samples collected in Slovenia included seventeen recreational waters (seawater, pool water, lake water, and river water; August 2004) and four wastewaters (January 2005). The most abundant UV filter was benzophenone-3 (11-400 ng L(-1)). Of the two anti-microbial agents studied, trace amounts, only, of triclosan were present in the river Kolpa (68 ng L(-1)) and in an hospital effluent (122 ng L(-1)).
Sigma-Aldrich
Benzyl cinnamate, 99%
Sigma-Aldrich
Bis(5-chloro-2-hydroxyphenyl)methane, 95%
Sigma-Aldrich
2-Benzyl-4-chlorophenol, 95%