MilliporeSigma
Nonenzymatic oligomerization of RNA by TNA templates.
Organic letters (2006-12-01)
Benjamin D Heuberger, Christopher Switzer
ABSTRACT
Cytosine TNA promotes nonenzymatic, template-directed oligomerization of complementary activated rGMP, leading to selective and efficient formation of RNA products. This process models "genetic takeover" of a pre-RNA by RNA. [reaction: see text]