MilliporeSigma
  • Utilization of hydrophobic bacterium Rhodococcus opacus B-4 as whole-cell catalyst in anhydrous organic solvents.
Utilization of hydrophobic bacterium Rhodococcus opacus B-4 as whole-cell catalyst in anhydrous organic solvents.
Applied microbiology and biotechnology (2006-11-24)
Shiho Yamashita, Masafumi Satoi, Yoshihiro Iwasa, Kohsuke Honda, Yuka Sameshima, Takeshi Omasa, Junichi Kato, Hisao Ohtake
ABSTRACT
Rhodococcus opacus strain B-4, which has recently been isolated as an organic solvent-tolerant bacterium, has a high hydrophobicity and exhibits a high affinity for hydrocarbons. This bacterium was able to survive for at least 5 days in organic solvents, including n-tetradecane, oleyl alcohol, and bis(2-ethylhexyl) phthalate (BEHP), which contained water less than 1% (w/v). The biocatalytic ability of R. opacus B-4 was demonstrated in the essentially nonaqueous BEHP using indigo production from indole as a model conversion. By the catabolism of oleic acid for NADH regeneration, indigo production increased up to 71.6 microg ml(-1) by 24 h.
Sigma-Aldrich
Oleyl alcohol, technical grade, 85%
Sigma-Aldrich
Oleyl alcohol, ≥99% (GC)