MilliporeSigma
  • Role of branched-chain fatty acids in pH stress tolerance in Listeria monocytogenes.
Role of branched-chain fatty acids in pH stress tolerance in Listeria monocytogenes.
Applied and environmental microbiology (2006-11-23)
Efstathios S Giotis, David A McDowell, Ian S Blair, Brian J Wilkinson
ABSTRACT
In alkaline conditions, Listeria monocytogenes cells develop higher proportions of branched-chain fatty acids (FAs), including more anteiso forms. In acid conditions, the opposite occurs. Reduced growth of pH-sensitive mutants at adverse pH (5.0/9.0) was alleviated by the addition of 2-methylbutyrate (an anteiso-FA precursor), suggesting that anteiso-FAs are important in adaptation to adverse pH. The balance between anteiso- and iso-FAs may be more important than changes in the amounts and/or degrees of saturation of FAs in pH adaptation.
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG