MilliporeSigma
TMSCH2Li-induced regioselective lithiation of (S)-nicotine.
Organic & biomolecular chemistry (2006-11-15)
Philippe C Gros, Abdelatif Doudouh, Christopher Woltermann
ABSTRACT
The first regioselective C-4 lithiation of (S)-nicotine has been realized using TMSCH2Li as basic reagent in toluene. The reaction proceeded under mild conditions with a small excess of electrophile. The 4-chloro derivative was subsequently metallated at C-5 with the same basic reagent in THF at -78 degrees C. This methodology opens a straightforward access to functional diversity in (S)-nicotine chemistry.
Product Description
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%