MilliporeSigma
  • Complexes of 2-thiophenecarbonyl and isonicotinoyl hydrazones of 3-(N-methyl)isatin. A study of their antimicrobial activity.
Complexes of 2-thiophenecarbonyl and isonicotinoyl hydrazones of 3-(N-methyl)isatin. A study of their antimicrobial activity.
Journal of inorganic biochemistry (2006-10-31)
María C Rodríguez-Argüelles, Sandra Mosquera-Vázquez, Patricia Tourón-Touceda, Jesús Sanmartín-Matalobos, Ana M García-Deibe, Marisa Belicchi-Ferrari, Giorgio Pelosi, Corrado Pelizzi, Franca Zani
ABSTRACT
Cobalt(II), nickel(II), copper(II) and zinc(II) complexes of 2-thiophenecarbonyl and isonicotinoyl hydrazones of 3-(N-methyl)isatin (HL(1) and HL(2), respectively) were synthesized and characterized, being the crystal structures of HL(1), HL(2) and [Ni(L(1))(2)].2CHCl(3) elucidated by X-ray diffraction techniques. The in vitro antimicrobial activity of all these compounds was tested against several bacteria and fungi. HL(1)and its complexes exhibited a strong inhibition of the growth of Haemophilus influenzae (MIC 0.15-1.50microg/mL) and good antibacterial properties towards Bacillus subtilis (MIC 3-25microg/mL). The minimal inhibitory concentration (MIC) was defined as the lowest concentration of compound inhibiting the growth of each strain. The antibacterial effectiveness was confirmed against a number of Gram positive bacteria, including methicillin-resistant Staphylococcus aureus. Yeasts and moulds showed a low susceptibility, except the dermatophyte mould Epidermophyton floccosum that is inhibited at concentrations ranging from 6 to 50microg/mL. In general, the antimicrobial activity of the thiophene derivatives was greater than that of the isonicotinic analogues.
Sigma-Aldrich
1-Methylisatin, 97%