MilliporeSigma
  • Beta-carotene and apocarotenals promote retinoid signaling in BEAS-2B human bronchioepithelial cells.
Beta-carotene and apocarotenals promote retinoid signaling in BEAS-2B human bronchioepithelial cells.
Archives of biochemistry and biophysics (2006-10-13)
Emmanuelle Kuntz, Ulrich Hoeller, Brad Greatrix, Christopher Lankin, Nicole Seifert, Samir Acharya, Georges Riss, Petra Buchwald-Hunziker, Willi Hunziker, Regina Goralczyk, Karin Wertz
ABSTRACT
High dose beta-carotene supplementation of smokers was associated with increased lung cancer risk in two intervention trials. It was proposed that generation of apocarotenals in smoke-exposed lungs impaired retinoic acid (RA) signaling, leading to squamous metaplasia and cell proliferation. To test this, we compared RA target gene regulation by retinoids, apocarotenals or beta-carotene by transcriptomics in BEAS-2B cells cultured to promote squamous differentiation. Retinoids, beta-carotene as well as apocarotenals induced known RA target genes. Retinoids upregulated involucrin, indicating that retinoids did not rescue BEAS-2B cells from squamous differentiation. Muc5AC, a marker for mucous differentiation, was transiently induced. beta-Carotene and apocarotenals less strongly induced involucrin and did not induce muc5AC. In summary, apocarotenals or beta-carotene upregulated RA target genes suggesting promotion, not inhibition, of RA signaling in BEAS-2B cells. Furthermore, apocarotenals and beta-carotene regulated gene expression independently of RA signaling. Squamous differentiation is not unequivocally linked to RA deficiency in BEAS-2B cells.
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ~20% apocarotenal basis (UV-vis), suspension (oily)
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ≥96.0% (UV)