MilliporeSigma
  • Formation of uniform single-crystalline bismuth sulfide nanowires under mixed-solvent condition.
Formation of uniform single-crystalline bismuth sulfide nanowires under mixed-solvent condition.
Journal of nanoscience and nanotechnology (2006-10-10)
Xiong Wang, Li Li, Shutao Wang, Qiaohan Guo, Zude Zhang, Yitai Qian
ABSTRACT
Uniform single-crystalline bismuth sulfide nanowires were prepared via a convenient solvothermal route under mixed-solvent condition. The nanowires were characterized by X-ray diffraction (XRD), high-resolution transmission electron microscopy (HRTEM), field-emission scanning electron microscopy (FESEM), energy-dispersive X-ray (EDX) spectrometry and UV-vis spectrophotometer. The growth mechanism of the one-dimensional nanostructures was discussed. The optical absorption band gap of the nanowires is about 1.7 eV, and the blue-shift phenomenon can be observed with respect of that of the bulk Bi2S3, which might bring in new types of applications.
Sigma-Aldrich
Bismuth(III) sulfide, 99%