MilliporeSigma
  • A green method for the electroorganic synthesis of new 1,3-Indandione derivatives.
A green method for the electroorganic synthesis of new 1,3-Indandione derivatives.
Chemical & pharmaceutical bulletin (2006-10-04)
Abdolmajid Bayandori Moghaddam, Mohammad Reza Ganjali, Parviz Norouzi, Maryam Latifi
ABSTRACT
This is an environmentally friendly method in the field of electroorganic reactions under controlled potential electrolysis, without toxic reagents at a carbon electrode in an undivided cell which involves the (EC) mechanism reaction and comprises two steps alternatively; (i) electrochemical oxidation and (ii) chemical reaction. In particular, the electrochemical oxidation of 4-tert-butylcatechol, 4-methylcatechol and 2,3-dihydroxybenzoic acid in the presence of 2-phenyl-1,3-indandione has been studied in a water-acetonitrile (90 : 10) mixture. The research includes the use of a variety of experimental techniques, such as cyclic voltammetry, controlled-potential electrolysis, and spectroscopic identification of products (FT-IR, (1)H-NMR, and MS spectrometry).
Sigma-Aldrich
1,3-Indandione, 97%