MilliporeSigma
  • Screening and characterization of variant Theta-class glutathione transferases catalyzing the activation of ethylene dibromide to a mutagen.
Screening and characterization of variant Theta-class glutathione transferases catalyzing the activation of ethylene dibromide to a mutagen.
Environmental and molecular mutagenesis (2006-09-02)
P David Josephy, Patricia L Taylor, Gaby Vervaet, Bengt Mannervik
ABSTRACT
Ethylene dibromide (EDB) is a widespread environmental pollutant and mutagen/carcinogen. Certain Theta-class glutathione transferases (GSTs), enzymes that catalyze the reaction of reduced glutathione (GSH) with electrophiles, activate EDB to a mutagen. Previous studies have shown that human GST T1-1, but not rat GST T2-2, activates EDB. We have constructed an E. coli lacZ reversion mutagenicity assay system in which expression of recombinant GST supports activation of EDB to a mutagen. Hexa-histidine N-terminal tagging of GST T1-1 results in greatly enhanced expression of the recombinant enzyme and gives a lacZ strain that shows a mutagenic response to EDB at extremely low levels (approximately 1 ng EDB per plate). The hexa-histidine-tagged enzyme was purified in one step by Ni(2+)-affinity chromatography. We applied the lacZ mutagenicity assay to the rapid screening of a library of variant GST Theta enzymes. Sequence variants with altered catalytic activities were identified, purified, and characterized.
Sigma-Aldrich
1,2-Dibromoethane, ≥99%
Sigma-Aldrich
1,2-Dibromoethane, purum, ≥98.0% (GC)
Sigma-Aldrich
1,2-Dibromoethane, 98%
Supelco
1,2-Dibromoethane solution, certified reference material, 200 μg/mL in methanol