MilliporeSigma
  • Reassignment of the configuration of salvianolic acid B and establishment of its identity with lithospermic acid B.
Reassignment of the configuration of salvianolic acid B and establishment of its identity with lithospermic acid B.
Journal of natural products (2006-08-29)
Anja Watzke, Steven J O'Malley, Robert G Bergman, Jonathan A Ellman
ABSTRACT
Salvianolic acid B and lithospermic acid B are the major components of Salvia miltiorrhiza, which is one of the most popular herbal traditional medicines in Asian countries. Salvianolic acid B and lithospermic acid B are reported to have identical structures except for the configurational assignments of two stereocenters. Through chemical correlation between a degradation product of salvianolic acid B and synthetic material, the absolute configuration of salvianolic acid B has been corrected to establish that salvianolic acid B and lithospermic acid B are in fact the same compound.
Sigma-Aldrich
Salvianolic acid B, ≥94% (HPLC)
Supelco
Salvianolic acid B, analytical standard